4-Methyl-1-phenyl-2-pentanol ≥96.0%

Supplier: TCI America

M2012-25G M2012-5G
TCM2012-25GEA 202.24 CAD
TCM2012-25G TCM2012-5G
4-Methyl-1-phenyl-2-pentanol ≥96.0%
4-Methyl-1-phenyl-2-pentanol

CAS Number: 7779-78-4
MDL Number: MFCD00209515
Molecular Formula: C12H18O
Molecular Weight: 178.28
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 123
Flash Point (°C): 110
Specific Gravity (20/20): 0.95

Formula: C₁₂H₁₈O
MW: 178.28 g/mol
Boiling Pt: 123 °C
Density: 0.95 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00209515
CAS Number: 7779-78-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca757617549a308c2a2d70d829e52d60145d1098&vid=ca757617549a308c2a2d70d829e52d60145d1098&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR