3-Methyl-1-phenyl-3-pentanol ≥98.0%
Supplier: TCI America
M2011-25G
M2011-5G
TCM2011-25GEA
60.04
CAD
TCM2011-25G
TCM2011-5G
3-Methyl-1-phenyl-3-pentanol ≥98.0%
3-Methyl-1-phenyl-3-pentanol
CAS Number: 10415-87-9
MDL Number: MFCD00021825
Molecular Formula: C12H18O
Molecular Weight: 178.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 113
Specific Gravity (20/20): 0.96
Formula:
C₁₂H₁₈O MW: 178.28 g/mol Boiling Pt: 130 °C Density: 0.96 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00021825 CAS Number: 10415-87-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...