2-Methyl-1-phenyl-2-butanol ≥98.0%

Supplier: TCI America

M2010-25G M2010-5G
TCM2010-25GEA 203.82 CAD
TCM2010-25G TCM2010-5G
2-Methyl-1-phenyl-2-butanol ≥98.0%
2-Methyl-1-phenyl-2-butanol

CAS Number: 772-46-3
MDL Number: MFCD01708473
Molecular Formula: C11H16O
Molecular Weight: 164.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 61
Specific Gravity (20/20): 0.97
Specific rotation [a]20/D: 0.9 deg

Formula: C₁₁H₁₆O
Boiling Pt: 61 °C
Density: 0.97 (20 °C)
MDL Number: MFCD01708473
CAS Number: 772-46-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87dc5e4de53a75c8d9ed0f906259c3fd0d626dae&vid=87dc5e4de53a75c8d9ed0f906259c3fd0d626dae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR