Methyl trans-4-(aminomethyl)cyclohexanecarboxylate hydrochloride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M1988-5G
M1988-25G
TCM1988-5GEA
161.16
CAD
TCM1988-5G
TCM1988-25G
Methyl trans-4-(aminomethyl)cyclohexanecarboxylate hydrochloride ≥98.0% (by GC, titration analysis)
Methyl trans-4-(aminomethyl)cyclohexanecarboxylate hydrochloride
CAS Number: 29275-88-5
MDL Number: MFCD00797465
Molecular Formula: C9H17NO2
Molecular Weight: 207.70
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Other Product Names / Synonyms: trans-4-(Aminomethyl)cyclohexanecarboxylic Acid Methyl Ester Hydrochloride
Formula:
C₉H₁₇NO₂ |
MDL Number:
MFCD00797465 CAS Number: 29275-88-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...