Methyl trans-4-aminocyclohexanecarboxylate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
Synonyms:
trans-4-Aminocyclohexanecarboxylic Acid Methyl Ester Hydrochloride
M1986-1G
M1986-5G
TCM1986-1GEA
143.78
CAD
TCM1986-1G
TCM1986-5G
Methyl trans-4-aminocyclohexanecarboxylate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Methyl trans-4-aminocyclohexanecarboxylate hydrochloride
CAS Number: 61367-07-5
MDL Number: MFCD08274538
Molecular Formula: C8H15NO2
Molecular Weight: 193.67
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Other Product Names / Synonyms: trans-4-Aminocyclohexanecarboxylic Acid Methyl Ester Hydrochloride
Formula:
C₈H₁₅NO₂ MW: 193.67 g/mol |
MDL Number:
MFCD08274538 CAS Number: 61367-07-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...