Methyl cis-4-aminocyclohexanecarboxylate hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: cis-Methyl 4-aminocyclohexanecarboxylate hydrochloride, cis-4-Aminocyclohexanecarboxylic Acid Methyl Ester Hydrochloride

M1985-5G M1985-1G
TCM1985-5GEA 216.46 CAD
TCM1985-5G TCM1985-1G
Methyl cis-4-aminocyclohexanecarboxylate hydrochloride ≥98.0%
Methyl cis-4-aminocyclohexanecarboxylate hydrochloride

CAS Number: 61367-16-6
MDL Number: MFCD08274537
Molecular Formula: C8H15NO2
Molecular Weight: 193.67
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 188


Other Product Names / Synonyms: cis-4-Aminocyclohexanecarboxylic Acid Methyl Ester Hydrochloride

Formula: C₈H₁₅NO₂
Melting Pt: 188 °C
MDL Number: MFCD08274537
CAS Number: 61367-16-6

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR