Methyl-5-(ethylsulfonyl)-2-methoxybenzoate ≥98.0%

Supplier: TCI America

Synonyms: 5-(Ethylsulfonyl)-2-methoxybenzoic Acid Methyl Ester, 5-(Ethylsulphonyl)-2-methoxybenzoic Acid Methyl Ester

M1975-25G M1975-5G
TCM1975-25GEA 434.66 CAD
TCM1975-25G TCM1975-5G
Methyl-5-(ethylsulfonyl)-2-methoxybenzoate ≥98.0%
Methyl-5-(ethylsulfonyl)-2-methoxybenzoate

CAS Number: 62140-67-4
MDL Number: MFCD01317547
Molecular Formula: C11H14O5S
Molecular Weight: 258.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 126


Other Product Names / Synonyms: 5-(Ethylsulfonyl)-2-methoxybenzoic Acid Methyl Ester

Formula: C₁₁H₁₄O₅S
MW: 258.29 g/mol
Melting Pt: 126 °C
MDL Number: MFCD01317547
CAS Number: 62140-67-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf1a4258ba46fd96af8f08467baa219bd26afe9f&vid=cf1a4258ba46fd96af8f08467baa219bd26afe9f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR