4-Methyl-2-pyridyl trifluoromethanesulfonate ≥97.0%

Supplier: TCI America

Synonyms: Trifluoromethanesulphonic acid 4-Methyl-2-pyridyl Ester, 4-Methyl-2-pyridyl Trifluoromethanesulfonate, 4-Methyl-2-pyridyl Trifluoromethanesulphonate, Trifluoromethanesulfonic acid 4-Methyl-2-pyridyl Ester

M1974-1G M1974-5G
TCM1974-1GEA 72.68 CAD
TCM1974-1G TCM1974-5G
4-Methyl-2-pyridyl trifluoromethanesulfonate ≥97.0%
4-Methyl-2-pyridyl trifluoromethanesulfonate

CAS Number: 179260-78-7
Molecular Formula: C7H6F3NO3S
Molecular Weight: 241.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.42
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: Trifluoromethanesulfonic Acid 4-Methyl-2-pyridyl Ester

Formula: C₇H₆F₃NO₃S
MW: 241.18 g/mol
Density: 1.42 (20 °C)
MDL Number: MFCD11042681
CAS Number: 179260-78-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e23998a7e2825c39f6fb0f1c07089f06bd51406&vid=9e23998a7e2825c39f6fb0f1c07089f06bd51406&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR