2,3-Anthracenedicarboxylic anhydride ≥93.0%

Supplier: TCI America

A1834-100MG A1834-500MG
TCA1834-100MGEA 164.32 CAD
TCA1834-100MG TCA1834-500MG
2,3-Anthracenedicarboxylic anhydride ≥93.0%
2,3-Anthracenedicarboxylic anhydride

CAS Number: 6812-14-2
Molecular Formula: C16H8O3
Molecular Weight: 248.24
Purity/Analysis Method: >93.0% (GC)
Form: Crystal

Formula: C₁₆H₈O₃
MW: 248.24 g/mol
CAS Number: 6812-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6290447d386efae60a7d0ca8666ce4d005923274&vid=6290447d386efae60a7d0ca8666ce4d005923274&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR