Methyl-β-D-ribofuranoside ≥97.0%
Supplier: TCI America
M1965-1G
M1965-5G
TCM1965-1GEA
75.84
CAD
TCM1965-1G
TCM1965-5G
Methyl-β-D-ribofuranoside ≥97.0%
Methyl-beta-D-ribofuranoside
CAS Number: 7473-45-2
MDL Number: MFCD00047075
Molecular Formula: C6H12O5
Molecular Weight: 164.16
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 78
Specific rotation [a]20/D: -51 deg (C=2, H2O)
Formula:
C₆H₁₂O₅ Melting Pt: 78 °C |
MDL Number:
MFCD00047075 CAS Number: 7473-45-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...