Methyl-2-methoxy-5-(methylsulfonyl)benzoate ≥98.0%

Supplier: TCI America

Synonyms: 2-Methoxy-5-(methylsulfonyl)benzoic acid Methyl Ester, Methyl 2-methoxy-5-(methylsulphonyl)benzoate, 2-Methoxy-5-(methylsulphonyl)benzoic acid Methyl Ester, Methyl 2-methoxy-5-(methylsulfonyl)benzoate

M1963-25G M1963-5G
TCM1963-25GEA 234.79 CAD
TCM1963-25G TCM1963-5G
Methyl-2-methoxy-5-(methylsulfonyl)benzoate ≥98.0%
Methyl-2-methoxy-5-(methylsulfonyl)benzoate

CAS Number: 63484-12-8
MDL Number: MFCD01317545
Molecular Formula: C10H12O5S
Molecular Weight: 244.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 117


Other Product Names / Synonyms: 2-Methoxy-5-(methylsulfonyl)benzoic Acid Methyl Ester

Formula: C₁₀H₁₂O₅S
Melting Pt: 117 °C
MDL Number: MFCD01317545
CAS Number: 63484-12-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7800437451879c025d638295832b933ced0218e4&vid=7800437451879c025d638295832b933ced0218e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR