ɑ-[2-(Methylamino)ethyl]benzyl alcohol ≥98.0%

Supplier: TCI America

Synonyms: 3-Hydroxy-N-methyl-3-phenylpropylamine, 3-(Methylamino)-1-phenylpropanol, α-[2-(Methylamino)ethyl]benzyl Alcohol, 3-(Methylamino)-1-phenylpropan-1-ol

M1960-25G M1960-5G
TCM1960-25GEA 165.9 CAD
TCM1960-25G TCM1960-5G
ɑ-[2-(Methylamino)ethyl]benzyl alcohol ≥98.0%
alpha-[2-(Methylamino)ethyl]benzyl alcohol

CAS Number: 42142-52-9
MDL Number: MFCD00674078
Molecular Formula: C10H15NO
Molecular Weight: 165.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 170
Melting point (°C): 64


Other Product Names / Synonyms: 3-Hydroxy-N-methyl-3-phenylpropylamine, 3-(Methylamino)-1-phenylpropanol

Formula: C₁₀H₁₅NO
Boiling Pt: 170 °C
Melting Pt: 64 °C
MDL Number: MFCD00674078
CAS Number: 42142-52-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77c822e306b160b1caaf6b70f308029d8dec3346&vid=77c822e306b160b1caaf6b70f308029d8dec3346&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR