N-Methylphenethylamine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

M1950-5G M1950-25G
TCM1950-5GEA 131.84 CAD
TCM1950-5G TCM1950-25G
N-Methylphenethylamine ≥97.0% (by GC, titration analysis)
N-Methylphenethylamine

CAS Number: 589-08-2
MDL Number: MFCD00008291
Molecular Formula: C9H13N
Molecular Weight: 135.21
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 73
Specific Gravity (20/20): 0.93


Other Product Names / Synonyms: N-Methylphenethylamine

Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 205 °C
Density: 0.93 (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00008291
CAS Number: 589-08-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b753c25305e438919fcc7472578ea617122b79a&vid=4b753c25305e438919fcc7472578ea617122b79a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR