trans-4-(4-Pentylcyclohexyl)benzonitrile ≥98.0%

Supplier: TCI America

Synonyms: 4-(trans-4-n-Pentylcyclohexyl)benzonitrile

A1828-25G A1828-5G
TCA1828-25GEA 395 CAD
TCA1828-25G TCA1828-5G
trans-4-(4-Pentylcyclohexyl)benzonitrile ≥98.0%
trans-4-(4-Pentylcyclohexyl)benzonitrile

CAS Number: 61204-01-1
MDL Number: MFCD00075548
Molecular Formula: C18H25N
Molecular Weight: 255.41
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 30
Flash Point (°C): 113


Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: 1-(trans-4-Amylcyclohexyl)-4-cyanobenzene, 4-(trans-4-Pentylcyclohexyl)benzonitrile

Formula: C₁₈H₂₅N
MW: 255.40 g/mol
Melting Pt: 30 °C
Flash Pt: 113 °C
MDL Number: MFCD00075548
CAS Number: 61204-01-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d77d1badec9b13c7dd1d8bd6abeb78f69a00b0c3&vid=d77d1badec9b13c7dd1d8bd6abeb78f69a00b0c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR