Methyl-2,3,6-tri-O-benzoyl-ɑ-D-galactopyranoside ≥98.0% (by HPLC)
Supplier: TCI America
M1933-250G
M1933-25G
M1933-5G
TCM1933-250GEA
1829.64
CAD
TCM1933-250G
TCM1933-25G
TCM1933-5G
Methyl-2,3,6-tri-O-benzoyl-ɑ-D-galactopyranoside ≥98.0% (by HPLC)
Methyl-2,3,6-tri-O-benzoyl-alpha-D-galactopyranoside
CAS Number: 3601-36-3
MDL Number: MFCD06200842
Molecular Formula: C28H26O9
Molecular Weight: 506.51
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 143
Specific rotation [a]20/D: 123 deg (C=1, CHCl3)
Storage Temperature: 0-10°C
Formula:
C₂₈H₂₆O₉ Melting Pt: 143 °C |
MDL Number:
MFCD06200842 CAS Number: 3601-36-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...