Methyl (4-piperidyl)acetate hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (4-Piperidyl)acetic Acid Methyl Ester Hydrochloride, Methyl 2-(piperidin-4-yl)acetate hydrochloride

M1926-1G M1926-5G
TCM1926-1GEA 31.6 CAD
TCM1926-1G TCM1926-5G
Methyl (4-piperidyl)acetate hydrochloride ≥98.0% (by titrimetric analysis)
Methyl (4-piperidyl)acetate hydrochloride

CAS Number: 81270-37-3
Molecular Formula: C8H15NO2
Molecular Weight: 193.67
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 155


Other Product Names / Synonyms: (4-Piperidyl)acetic Acid Methyl Ester Hydrochloride

Formula: C₈H₁₅NO₂
MW: 193.67 g/mol
Melting Pt: 155 °C
MDL Number: MFCD09261419
CAS Number: 81270-37-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87d9b96d02ea6f11a36e3607a3cce30ad506dcbc&vid=87d9b96d02ea6f11a36e3607a3cce30ad506dcbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR