1-Methyl-3-octylimidazolium bromide ≥98.0%
Supplier: TCI America
Synonyms:
1-Methyl-3-octylimidazolium Bromide
M1904-25G
M1904-5G
TCM1904-25GEA
508.76
CAD
TCM1904-25G
TCM1904-5G
1-Methyl-3-octylimidazolium bromide ≥98.0%
1-Methyl-3-octylimidazolium bromide
CAS Number: 61545-99-1
Molecular Formula: C12H23BrN2
Molecular Weight: 275.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Clear Liquid
Formula:
C₁₂H₂₃BrN₂ MW: 275.23 g/mol |
MDL Number:
MFCD10566918 CAS Number: 61545-99-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...