4-Methyl-N,N-diphenylaniline ≥98.0%

Supplier: TCI America

M1898-1G M1898-5G
TCM1898-1GEA 178.54 CAD
TCM1898-1G TCM1898-5G
4-Methyl-N,N-diphenylaniline ≥98.0%
4-Methyl-N,N-diphenylaniline

CAS Number: 4316-53-4
Molecular Formula: C19H17N
Molecular Weight: 259.35
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 67


Other Product Names / Synonyms: N,N-Diphenyl-p-toluidine

Formula: C₁₉H₁₇N
Melting Pt: 67 °C
MDL Number: MFCD03093256
CAS Number: 4316-53-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aadc245b00a1d9992cbe43bda7264fae678e96c1&vid=aadc245b00a1d9992cbe43bda7264fae678e96c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR