4,4'-Methylenebis(2,6-diethylaniline) ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Bis(4-amino-3,5-diethylphenyl)methane, 4,4-Methylenebis(2,6-diethylaniline), 4,4'-Diamino-3,3',5,5'-tetraethyldiphenylmethane

M1897-500G M1897-25G
TCM1897-500GEA 417.12 CAD
TCM1897-500G TCM1897-25G
4,4'-Methylenebis(2,6-diethylaniline) ≥98.0% (by GC, titration analysis)
4,4'-Methylenebis(2,6-diethylaniline)

CAS Number: 13680-35-8
MDL Number: MFCD00071552
Molecular Formula: C21H30N2
Molecular Weight: 310.49
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 205
Melting point (°C): 90


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: Bis(4-amino-3,5-diethylphenyl)methane, 4,4'-Diamino-3,3',5,5'-tetraethyldiphenylmethane

Formula: C₂₁H₃₀N₂
MW: 310.4763 g/mol
Boiling Pt: 205 °C
Melting Pt: 90 °C
MDL Number: MFCD00071552
CAS Number: 13680-35-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06c76b74f09da306749d1d11373aabb3aa1c8f53&vid=06c76b74f09da306749d1d11373aabb3aa1c8f53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR