Methyl-1,2,3,4-tetra-O-acetyl-β-D-glucuronate ≥96.0%
Supplier: TCI America
CAS Number: 7355-18-2
MDL Number: MFCD00069834
Molecular Formula: C15H20O11
Molecular Weight: 376.31
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 179
Specific rotation [a]20/D: 8 deg (C=1, CHCl3)
Product Categories: Bioscience, Glucuronic Acids, O-Substituted Sugars
Other Product Names / Synonyms: 1,2,3,4-Tetra-O-acetyl-beta-D-glucuronic Acid Methyl Ester, Methyl 1,2,3,4-Tetra-O-acetyl-beta-D-glucopyranuronate, 1,2,3,4-Tetra-O-acetyl-beta-D-glucopyranuronic Acid Methyl Ester
Formula:
C₁₅H₂₀O₁₁ Melting Pt: 179 °C |
MDL Number:
MFCD00069834 CAS Number: 7355-18-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...