Methyl-2-oxovalerate ≥90.0%

Supplier: TCI America

M1863-5G M1863-25G
TCM1863-5GEA 470.84 CAD
TCM1863-5G TCM1863-25G
Methyl-2-oxovalerate ≥90.0%
Methyl-2-oxovalerate

CAS Number: 6376-59-6
Molecular Formula: C6H10O3
Molecular Weight: 130.14
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 62
Specific Gravity (20/20): 1.03
Storage Temperature: 0-10°C


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: 2-Oxovaleric Acid Methyl Ester, Methyl 2-Oxopentanoate, 2-Oxopentanoic Acid Methyl Ester

Formula: C₆H₁₀O₃
Boiling Pt: 62 °C
Density: 1.03 (20 °C)
MDL Number: MFCD10566912
CAS Number: 6376-59-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=617522eb6d19e74a752739fed43609c0ac8ae899&vid=617522eb6d19e74a752739fed43609c0ac8ae899&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR