1-Methyl-L-aspartate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M1859-1G M1859-5G
TCM1859-1GEA 41.08 CAD
TCM1859-1G TCM1859-5G
1-Methyl-L-aspartate ≥98.0% (by titrimetric analysis)
1-Methyl-L-aspartate

CAS Number: 17812-32-7
MDL Number: MFCD00080289
Molecular Formula: C5H9NO4
Molecular Weight: 147.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 186
Specific rotation [a]20/D: 36 deg (C=0.5, H2O)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: L-Aspartic Acid 1-Methyl Ester, H-Asp-OMe

Formula: C₅H₉NO₄
Melting Pt: 186 °C
MDL Number: MFCD00080289
CAS Number: 17812-32-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6116583aca8d3c49c646b842db379c3d18fc7a05&vid=6116583aca8d3c49c646b842db379c3d18fc7a05&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR