4-Methoxythioanisole ≥98.0%
Supplier: TCI America
M1852-5G
M1852-25G
TCM1852-5GEA
55.3
CAD
TCM1852-5G
TCM1852-25G
4-Methoxythioanisole ≥98.0%
4-Methoxythioanisole
CAS Number: 1879-16-9
MDL Number: MFCD00010587
Molecular Formula: C8H10OS
Molecular Weight: 154.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Flash Point (°C): 108
Specific Gravity (20/20): 1.11
Other Product Names / Synonyms: 4-(Methylthio)anisole
Formula:
C₈H₁₀OS MW: 154.2294 g/mol Boiling Pt: 137 °C Density: 1.11 (20 °C) Flash Pt: 108 °C |
MDL Number:
MFCD00010587 CAS Number: 1879-16-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...