2-Methoxythioanisole ≥98.0%

Supplier: TCI America

M1851-5G M1851-25G
TCM1851-5GEA 53.72 CAD
TCM1851-5G TCM1851-25G
2-Methoxythioanisole ≥98.0%
2-Methoxythioanisole

CAS Number: 2388-73-0
MDL Number: MFCD00792424
Molecular Formula: C8H10OS
Molecular Weight: 154.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 142
Melting point (°C): 19
Flash Point (°C): 110
Specific Gravity (20/20): 1.14


Other Product Names / Synonyms: 2-(Methylthio)anisole

Formula: C₈H₁₀OS
Boiling Pt: 142 °C
Melting Pt: 19 °C
Density: 1.14 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00792424
CAS Number: 2388-73-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56aa7c2687651566c5bd14bb99d8e27dcdc1f6ae&vid=56aa7c2687651566c5bd14bb99d8e27dcdc1f6ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR