Methyl-4-chloropicolinate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 4-chloropicolinate, Methyl 4-chloropyridine-2-carboxylate

M1832-1G M1832-5G
TCM1832-1GEA 33.18 CAD
TCM1832-1G TCM1832-5G
Methyl-4-chloropicolinate ≥98.0%
Methyl 4-chloropicolinate

CAS Number: 24484-93-3
MDL Number: MFCD04116183
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 59


Other Product Names / Synonyms: 4-Chloropicolinic Acid Methyl Ester, Methyl 4-Chloropicolinate, 4-Chloro-2-pyridinecarboxylic Acid Methyl Ester

Formula: C₇H₆ClNO₂
MW: 171.5800 g/mol
Melting Pt: 59 °C
MDL Number: MFCD04116183
CAS Number: 24484-93-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb887dc521dd803d391773267a3f4dd6ac2ac072&vid=bb887dc521dd803d391773267a3f4dd6ac2ac072&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR