Mono(2-acryloyloxyethyl) succinate ≥90.0% stabilized
Supplier: TCI America
Synonyms:
2-(Acryloyloxy)ethyl Hydrogen Succinate, 4-(2-(Acryloyloxy)ethoxy)-4-oxobutanoic acid, Succinic Acid Mono(2-acryloyloxyethyl) Ester
(stabilized with MEHQ)
CAS Number: 50940-49-3
MDL Number: MFCD00274301
Molecular Formula: C9H12O6
Molecular Weight: 216.19
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Flash Point (°C): 178
Specific Gravity (20/20): 1.23
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Succinic Acid Mono(2-acryloyloxyethyl) Ester, 2-(Acryloyloxy)ethyl Hydrogen Succinate
Formula:
C₉H₁₂O₆ MW: 216.19 g/mol Density: 1.23 (20 °C) Flash Pt: 178 °C |
MDL Number:
MFCD00274301 CAS Number: 50940-49-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...