Methyl-2-(p-tolyl)benzoate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 2-(4-Methylphenyl)benzoate, 4'-Methylbiphenyl-2-carboxylic Acid Methyl Ester, Methyl 4'-methyl-[1,1'-biphenyl]-2-carboxylate, Methyl 4'-Methylbiphenyl-2-carboxylate, 2-(p-Tolyl)benzoic Acid Methyl Ester

M1809-5G M1809-25G
TCM1809-5GEA 61.62 CAD
TCM1809-5G TCM1809-25G
Methyl-2-(p-tolyl)benzoate ≥98.0%
Methyl-2-(p-tolyl)benzoate

CAS Number: 114772-34-8
MDL Number: MFCD03453661
Molecular Formula: C15H14O2
Molecular Weight: 226.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 59


Other Product Names / Synonyms: 2-(p-Tolyl)benzoic Acid Methyl Ester, Methyl 2-(4-Methylphenyl)benzoate, Methyl 4'-Methylbiphenyl-2-carboxylate, 4'-Methylbiphenyl-2-carboxylic Acid Methyl Ester

Formula: C₁₅H₁₄O₂
MW: 226.28 g/mol
Melting Pt: 59 °C
MDL Number: MFCD03453661
CAS Number: 114772-34-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cde57e1489f87c2ecbfba72bc5a5ebc54b58244&vid=7cde57e1489f87c2ecbfba72bc5a5ebc54b58244&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR