2-[4-(Bromomethyl)phenyl]benzoate ≥98.0%
Supplier: TCI America
Synonyms:
Methyl 2-[4-(Bromomethyl)phenyl]benzoate
M1806-5G
M1806-25G
TCM1806-5GEA
58.46
CAD
TCM1806-5G
TCM1806-25G
2-[4-(Bromomethyl)phenyl]benzoate ≥98.0%
2-[4-(Bromomethyl)phenyl]benzoate
CAS Number: 114772-38-2
MDL Number: MFCD06200816
Molecular Formula: C15H13BrO2
Molecular Weight: 305.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 56
Other Product Names / Synonyms: 2-[4-(Bromomethyl)phenyl]benzoic Acid Methyl Ester, Methyl 4'-(Bromomethyl)biphenyl-2-carboxylate, 4'-(Bromomethyl)biphenyl-2-carboxylic Acid Methyl Ester
Formula:
C₁₅H₁₃BrO₂ MW: 305.1665 g/mol Melting Pt: 56 °C |
MDL Number:
MFCD06200816 CAS Number: 114772-38-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...