6-Methoxy-o-toluidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M1804-25G M1804-5G
TCM1804-25GEA 635.16 CAD
TCM1804-25G TCM1804-5G
6-Methoxy-o-toluidine ≥98.0% (by GC, titration analysis)
6-Methoxy-o-toluidine

CAS Number: 50868-73-0
MDL Number: MFCD00075455
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 116
Flash Point (°C): 113
Freezing point (°C): 28


Other Product Names / Synonyms: 6-Methoxy-o-toluidine, 2-Amino-1-methoxy-3-methylbenzene

Formula: C₈H₁₁NO
Boiling Pt: 116 °C
Flash Pt: 113 °C
MDL Number: MFCD00075455
CAS Number: 50868-73-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=203fd49e9d6253a19747854a7643bfe27e0cf52c&vid=203fd49e9d6253a19747854a7643bfe27e0cf52c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR