Mefenamic acid ≥98.0%

Supplier: TCI America

Synonyms: 2-(2,3-Xylidino)benzoic acid, 2-((2,3-Dimethylphenyl)amino)benzoic acid, 2-(2,3-Dimethylphenylamino)benzoic acid, N-(2,3-Dimethylphenyl)anthranilic acid

M1782-500G M1782-25G M1782-100G
TCM1782-500GEA 392.16 CAD
TCM1782-500G TCM1782-25G TCM1782-100G
Mefenamic acid ≥98.0%
Mefenamic acid

CAS Number: 61-68-7
MDL Number: MFCD00051721
Molecular Formula: C15H15NO2
Molecular Weight: 241.29
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 230


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Anti-rheumatoid Arthritis Ingredients
Other Product Names / Synonyms: N-(2,3-Dimethylphenyl)anthranilic Acid, 2-(2,3-Dimethylphenylamino)benzoic Acid, 2-(2,3-Xylidino)benzoic Acid

Formula: C₁₅H₁₅NO₂
MW: 241.29 g/mol
Melting Pt: 230 °C
MDL Number: MFCD00051721
CAS Number: 61-68-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5345375d041702cf922fb762285171bea8fded72&vid=5345375d041702cf922fb762285171bea8fded72&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR