Methyl-3-amino-4-chlorobenzoate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Methyl 3-amino-4-chlorobenzoate, Methyl 3-Amino-4-chlorobenzoate, 3-amino-4-chlorobenzoic acid methylester

M1778-5G M1778-25G
TCM1778-5GEA 86.9 CAD
TCM1778-5G TCM1778-25G
Methyl-3-amino-4-chlorobenzoate ≥98.0% (by GC, titration analysis)
Methyl 3-amino-4-chlorobenzoate

CAS Number: 40872-87-5
Molecular Formula: C8H8ClNO2
Molecular Weight: 185.61
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 83


Other Product Names / Synonyms: 3-Amino-4-chlorobenzoic Acid Methyl Ester

Formula: C₈H₈ClNO₂
MW: 185.6100 g/mol
Melting Pt: 83 °C
MDL Number: MFCD03470533
CAS Number: 40872-87-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c167552700f3e7509ef261caa8dec0bbcb48a2c1&vid=c167552700f3e7509ef261caa8dec0bbcb48a2c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR