LacDiNAc(I) MP glycoside
Supplier: TCI America
Synonyms:
4-Methoxyphenyl 3-O-(2-Acetamido-2-deoxy-beta-D-galactopyranosyl)-2-acetamido-2-deoxy-beta-D-glucopyranoside
M1776-5MG
TCM1776-5MGEA
1101.96
CAD
TCM1776-5MG
LacDiNAc(I) MP glycoside
LacDiNAc(I) MP glycoside
Molecular Weight: 530.53
Form: Crystal
Color: White
Storage Temperature: <0°C
Molecular Formula: C23H34N2O12
Other Product Names / Synonyms: 4-Methoxyphenyl 3-O-(2-Acetamido-2-deoxy-beta-D-galactopyranosyl)-2-acetamido-2-deoxy-beta-D-glucopyranoside
Formula:
C₂₃H₃₄N₂O₁₂ MW: 530.53 g/mol |
CAS Number:
1858223-95-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...