2-Methyl-1-naphthyl acetate ≥98.0%

Supplier: TCI America

M1757-25G M1757-250G
TCM1757-25GEA 50.56 CAD
TCM1757-25G TCM1757-250G
2-Methyl-1-naphthyl acetate ≥98.0%
2-Methyl-1-naphthyl acetate

CAS Number: 5697-02-9
Molecular Formula: C13H12O2
Molecular Weight: 200.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 82


Other Product Names / Synonyms: Acetic Acid 2-Methyl-1-naphthyl Ester, 1-Acetoxy-2-methylnaphthalene

Formula: C₁₃H₁₂O₂
Melting Pt: 82 °C
CAS Number: 5697-02-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e62f5b6f4532b69b15f9198139bdc6f3bc98d086&vid=e62f5b6f4532b69b15f9198139bdc6f3bc98d086&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR