4-Methoxy-N-methylaniline ≥96.0%
Supplier: TCI America
Synonyms:
N-Methyl-p-anisidine
M1743-5G
M1743-1G
TCM1743-5GEA
132.72
CAD
TCM1743-5G
TCM1743-1G
4-Methoxy-N-methylaniline ≥96.0%
4-Methoxy-N-methylaniline
CAS Number: 5961-59-1
MDL Number: MFCD00008399
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Boiling point (°C): 136
Melting point (°C): 35
Flash Point (°C): 110
Other Product Names / Synonyms: 4-Methoxy-N-methylaniline
Formula:
C₈H₁₁NO Boiling Pt: 136 °C Melting Pt: 35 °C Flash Pt: 110 °C |
MDL Number:
MFCD00008399 CAS Number: 5961-59-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...