4'-Methoxy-N-methylformanilide ≥97.0%

Supplier: TCI America

M1742-1G M1742-5G
TCM1742-1GEA 257.86 CAD
TCM1742-1G TCM1742-5G
4'-Methoxy-N-methylformanilide ≥97.0%
4'-Methoxy-N-methylformanilide

CAS Number: 5279-51-6
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Freezing point (°C): 26


Other Product Names / Synonyms: N-(4-Methoxyphenyl)-N-methylformamide

Formula: C₉H₁₁NO₂
MDL Number: MFCD09038534
CAS Number: 5279-51-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b2e3fcffcb7b5a7eb02f7f4bfa1d7c8488eb638&vid=7b2e3fcffcb7b5a7eb02f7f4bfa1d7c8488eb638&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR