Methyl-2,3-dihydroxybenzoate ≥98.0%
Supplier: TCI America
M1737-25G
M1737-5G
TCM1737-25GEA
277.76
CAD
TCM1737-25G
TCM1737-5G
Methyl-2,3-dihydroxybenzoate ≥98.0%
Methyl 2,3-dihydroxybenzoate
CAS Number: 2411-83-8
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 96
Melting point (°C): 81
Other Product Names / Synonyms: 2,3-Dihydroxybenzoic Acid Methyl Ester
Formula:
C₈H₈O₄ Boiling Pt: 96 °C Melting Pt: 81 °C |
MDL Number:
MFCD00039520 CAS Number: 2411-83-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...