2-Acetamido-4,6-O-benzylidene-2-deoxy-β-D-glucopyranosyl azide ≥98.0%

Supplier: TCI America

Synonyms: 2-​Acetamido-​4,​6-​O-​benzylidene-​2-​deoxy-​β-​D-​glucopyranosyl azide

A1811-5G A1811-1G
TCA1811-5GEA 1213.44 CAD
TCA1811-5G TCA1811-1G
2-Acetamido-4,6-O-benzylidene-2-deoxy-β-D-glucopyranosyl azide ≥98.0%
2-Acetamido-4,6-O-benzylidene-2-deoxy-beta-D-glucopyranosyl azide

CAS Number: 168397-51-1
Molecular Formula: C15H18N4O5
Molecular Weight: 334.33
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Storage Temperature: <0°C

Formula: C₁₅H₁₈N₄O₅
MW: 334.33 g/mol
CAS Number: 168397-51-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa998856c6893162cef0dd40e7e87804dc7d211e&vid=aa998856c6893162cef0dd40e7e87804dc7d211e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR