LacDiNAc MP glycoside
Supplier: TCI America
Synonyms:
4-Methoxyphenyl 4-O-(2-Acetamido-2-deoxy-beta-D-galactopyranosyl)-2-acetamido-2-deoxy-beta-D-glucopyranoside
M1733-5MG
TCM1733-5MGEA
1769.54
CAD
TCM1733-5MG
LacDiNAc MP glycoside
LacDiNAc MP glycoside
Molecular Weight: 530.53
Storage Temperature: <0°C
Molecular Formula: C23H34N2O12
Other Product Names / Synonyms: 4-Methoxyphenyl 4-O-(2-Acetamido-2-deoxy-beta-D-galactopyranosyl)-2-acetamido-2-deoxy-beta-D-glucopyranoside
Formula:
C₂₃H₃₄N₂O₁₂ MW: 530.53 g/mol |
CAS Number:
1858224-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...