Gal[236Bn]β(1-4)Glc[236Bn]-β-MP ≥92.0% (by HPLC)
Supplier: TCI America
M1726-1G
M1726-5G
TCM1726-1GEA
406.69
CAD
TCM1726-1G
TCM1726-5G
Gal[236Bn]β(1-4)Glc[236Bn]-β-MP ≥92.0% (by HPLC)
Gal[236Bn]beta(1-4)Glc[236Bn]-beta-MP
CAS Number: 150412-81-0
Molecular Formula: C61H64O12
Molecular Weight: 989.17
Purity/Analysis Method: >92.0% (HPLC)
Form: Crystal
Storage Temperature: <0°C
Product Categories: Synthetic Organic Chemistry, Glycosylation, Glycosylation, Bioscience, Disaccharides, Glycosides, p-Methoxyphenyl Glycosides, O-Substituted Sugars
Other Product Names / Synonyms: LacMP 4'-OH, per OBn, 4-Methoxyphenyl 4-O-(2,3,6-Tri-O-benzyl-beta-D-galactopyranosyl)-2,3,6-tri-O-benzyl-beta-D-glucopyranoside
Formula:
C₆₁H₆₄O₁₂ |
CAS Number:
150412-81-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...