Methyl quinoline-6-carboxylate ≥97.0% (by GC, titration analysis)

Supplier: TCI America

M1687-5G M1687-25G
TCM1687-5GEA 75.84 CAD
TCM1687-5G TCM1687-25G
Methyl quinoline-6-carboxylate ≥97.0% (by GC, titration analysis)
Methyl quinoline-6-carboxylate

CAS Number: 38896-30-9
Molecular Formula: C11H9NO2
Molecular Weight: 187.20
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 85


Other Product Names / Synonyms: 6-Quinolinecarboxylic Acid Methyl Ester

Formula: C₁₁H₉NO₂
Melting Pt: 85 °C
MDL Number: MFCD00956766
CAS Number: 38896-30-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8d149942c96f4d574de65e8281b1fc4dd2e5cac&vid=b8d149942c96f4d574de65e8281b1fc4dd2e5cac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR