Methyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside ≥98.0%

Supplier: TCI America

M1682-1G M1682-5G
TCM1682-1GEA 255.96 CAD
TCM1682-1G TCM1682-5G
Methyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside ≥98.0%
Methyl-2,3,4,6-tetra-O-acetyl-1-thio-beta-D-glucopyranoside

CAS Number: 13350-45-3
Molecular Formula: C15H22O9S
Molecular Weight: 378.39
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 91
Specific rotation [a]20/D: -15 deg (C=1, CHCl3)
Storage Temperature: 0-10°C

Formula: C₁₅H₂₂O₉S
Melting Pt: 91 °C
CAS Number: 13350-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4cf90cf59bc77b5b831e87fa82710c1e47208a7&vid=e4cf90cf59bc77b5b831e87fa82710c1e47208a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR