2-Methyl-5-nitroanisole ≥98.0%
Supplier: TCI America
|
Warning
|
M1681-25G
M1681-500G
TCM1681-25GEA
41.08
CAD
TCM1681-25G
TCM1681-500G
2-Methyl-5-nitroanisole ≥98.0%
2-Methyl-5-nitroanisole
CAS Number: 13120-77-9
Molecular Formula: C8H9NO3
Molecular Weight: 167.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 73
Other Product Names / Synonyms: 1-Methoxy-2-methyl-5-nitrobenzene, 2-Methyl-5-nitroanisole
Formula:
C₈H₉NO₃ Melting Pt: 73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043912 CAS Number: 13120-77-9 EINECS: 236-048-6 UN: 2446 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...