4'-(Methylsulfonyl)acetophenone ≥98.0%

Supplier: TCI America

M1678-250G M1678-25G
TCM1678-250GEA 776.89 CAD
TCM1678-250G TCM1678-25G
4'-(Methylsulfonyl)acetophenone ≥98.0%
4'-(Methylsulphonyl)acetophenone

CAS Number: 10297-73-1
Molecular Formula: C9H10O3S
Molecular Weight: 198.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 129

Formula: C₉H₁₀O₃S
Melting Pt: 129 °C
MDL Number: MFCD00025072
CAS Number: 10297-73-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d687dabd9e688d4061ede93248e5b2b43c24013&vid=0d687dabd9e688d4061ede93248e5b2b43c24013&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR