7-Methyl-1,4-benzothiazin-3-one

Supplier: TCI America

M1661-5G M1661-1G
TCM1661-5GEA 160.37 CAD
TCM1661-5G TCM1661-1G
7-Methyl-1,4-benzothiazin-3-one
7-Methyl-1,4-benzothiazin-3-one

CAS Number: 30322-02-2
Molecular Formula: C9H9NOS
Molecular Weight: 179.24
Form: Crystal
Color: White
Melting point (°C): 206

Formula: C₉H₉NOS
Melting Pt: 206 °C
MDL Number: MFCD02660690
CAS Number: 30322-02-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3572d7f280c7d431f57f3898ee88a2baeae54c0&vid=d3572d7f280c7d431f57f3898ee88a2baeae54c0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR