4-(3-Aminopropyl)thiomorpholine-1,1-dioxide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-(1,1-Dioxothiomorpholino)propylamine

A1804-5G A1804-1G
TCA1804-5GEA 246.32 CAD
TCA1804-5G TCA1804-1G
4-(3-Aminopropyl)thiomorpholine-1,1-dioxide ≥98.0% (by GC, titration analysis)
4-(3-Aminopropyl)thiomorpholine-1,1-dioxide

CAS Number: 90000-25-2
Molecular Formula: C7H16N2O2S
Molecular Weight: 192.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 190
Freezing point (°C): 31


Other Product Names / Synonyms: 3-(1,1-Dioxothiomorpholino)propylamine

Formula: C₇H₁₆N₂O₂S
MW: 192.28 g/mol
Boiling Pt: 190 °C
CAS Number: 90000-25-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c975a094b008ee43f4dda5bce016c2a4fc00f6c&vid=4c975a094b008ee43f4dda5bce016c2a4fc00f6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR