Methyl-3,4,6-tri-O-acetyl-2-deoxy-2-phthalimido-1-thio-β-D-glucopyranoside ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

M1649-5G M1649-1G
TCM1649-5GEA 962.06 CAD
TCM1649-5G TCM1649-1G
Methyl-3,4,6-tri-O-acetyl-2-deoxy-2-phthalimido-1-thio-β-D-glucopyranoside ≥98.0% (by HPLC, total nitrogen)
Methyl-3,4,6-tri-O-acetyl-2-deoxy-2-phthalimido-1-thio-beta-D-glucopyranoside

CAS Number: 79528-48-6
Molecular Formula: C21H23NO9S
Molecular Weight: 465.47
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 150
Specific rotation [a]20/D: 49.5 deg (C=1, CHCl3)
Storage Temperature: <0°C

Formula: C₂₁H₂₃NO₉S
MW: 465.47 g/mol
Melting Pt: 150 °C
CAS Number: 79528-48-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e313068d0191b84e43fe2878df05ce58db86673&vid=4e313068d0191b84e43fe2878df05ce58db86673&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR