4-Methoxyphenyl-2,4,6-tri-O-acetyl-3-O-benzyl-β-D-glucopyranoside ≥96.0%

Supplier: TCI America

M1642-1G M1642-5G
TCM1642-1GEA 271.13 CAD
TCM1642-1G TCM1642-5G
4-Methoxyphenyl-2,4,6-tri-O-acetyl-3-O-benzyl-β-D-glucopyranoside ≥96.0%
4-Methoxyphenyl-2,4,6-tri-O-acetyl-3-O-benzyl-beta-D-glucopyranoside

CAS Number: 303127-79-9
Molecular Formula: C26H30O10
Molecular Weight: 502.52
Purity/Analysis Method: >96.0% (HPLC)
Form: Crystal
Melting point (°C): 160
Specific rotation [a]20/D: -22 deg (C=1, CHCl3)
Storage Temperature: 0-10°C

Formula: C₂₆H₃₀O₁₀
MW: 502.52 g/mol
Melting Pt: 160 °C
CAS Number: 303127-79-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e85d8e70e7aba84b51952f2fbdb172ce69a8da69&vid=e85d8e70e7aba84b51952f2fbdb172ce69a8da69&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR