4-Methoxyphenyl-3-O-benzyl-4,6-O-benzylidene-β-D-glucopyranoside ≥98.0% (by HPLC)

Supplier: TCI America

M1640-1G M1640-5G
TCM1640-1GEA 628.05 CAD
TCM1640-1G TCM1640-5G
4-Methoxyphenyl-3-O-benzyl-4,6-O-benzylidene-β-D-glucopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-3-O-benzyl-4,6-O-benzylidene-beta-D-glucopyranoside

CAS Number: 303127-81-3
Molecular Formula: C27H28O7
Molecular Weight: 464.51
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 207
Specific rotation [a]20/D: -20 deg (C=1, CHCl3)
Storage Temperature: <0°C

Formula: C₂₇H₂₈O₇
Melting Pt: 207 °C
CAS Number: 303127-81-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57ccfb377271f4514ec185eac8e867a035743ef4&vid=57ccfb377271f4514ec185eac8e867a035743ef4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR