4-Methylbenzyl phenyl ketone ≥97.0%

Supplier: TCI America

M1639-25G M1639-5G
TCM1639-25GEA 377.62 CAD
TCM1639-25G TCM1639-5G
4-Methylbenzyl phenyl ketone ≥97.0%
4-Methylbenzyl phenyl ketone

CAS Number: 2430-99-1
Molecular Formula: C15H14O
Molecular Weight: 210.28
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 155
Melting point (°C): 95


Other Product Names / Synonyms: 2-(p-Tolyl)acetophenone, p-Tolylmethyl Phenyl Ketone

Formula: C₁₅H₁₄O
Boiling Pt: 155 °C
Melting Pt: 95 °C
MDL Number: MFCD00017513
CAS Number: 2430-99-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd458710204b95c639c2386c2ba4b517d8d3e91c&vid=cd458710204b95c639c2386c2ba4b517d8d3e91c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR