4-(2-Aminoethyl)thiomorpholine-1,1-dioxide ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(1,1-Dioxothiomorpholino)ethylamine

A1803-1G A1803-5G
TCA1803-1GEA 206.98 CAD
TCA1803-1G TCA1803-5G
4-(2-Aminoethyl)thiomorpholine-1,1-dioxide ≥97.0% (by GC, titration analysis)
4-(2-Aminoethyl)thiomorpholine-1,1-dioxide

CAS Number: 89937-52-0
Molecular Formula: C6H14N2O2S
Molecular Weight: 178.25
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 182
Melting point (°C): 92


Other Product Names / Synonyms: 2-(1,1-Dioxothiomorpholino)ethylamine

Formula: C₆H₁₄N₂O₂S
MW: 178.25 g/mol
Boiling Pt: 182 °C
Melting Pt: 92 °C
CAS Number: 89937-52-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=122cc41b0dc0766b899102178d70d0d8078bc7cf&vid=122cc41b0dc0766b899102178d70d0d8078bc7cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR